missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylaniline, 99%, Thermo Scientific Chemicals

24.82 € - 385.92 €

Chemical Identifiers

CAS 121-69-7
Molecular Formula C8H11N
Molecular Weight (g/mol) 121.18
MDL Number MFCD00008304
InChI Key JLTDJTHDQAWBAV-UHFFFAOYSA-N
Synonym dimethylaniline, dimethylphenylamine, n,n-dimethylbenzenamine, benzenamine, n,n-dimethyl, dimethylamino benzene, n,n-dimethylphenylamine, n,n-dimethylbenzeneamine, dimethylaminobenzene, dwumetyloanilina, n,n-dimethylamino benzene
PubChem CID 949
ChEBI CHEBI:16269
IUPAC Name N,N-dimethylaniline
SMILES CN(C)C1=CC=CC=C1
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11488770
View Documents
Thermo Scientific Alfa Aesar
A11916.AE
100 mL
24.82 €
100mL
Please to purchase this item. Need a web account? Register with us today!
11483894
View Documents
Thermo Scientific Alfa Aesar
A11916.AP
500 mL
36.88 €
500mL
Please to purchase this item. Need a web account? Register with us today!
11493894
View Documents
Thermo Scientific Alfa Aesar
A11916.0F
2500 mL
119.60 €
2500mL
Please to purchase this item. Need a web account? Register with us today!
11403904
View Documents
Thermo Scientific Alfa Aesar
A11916.0D
10,000 mL
385.92 €
10000mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

N,N-Dimethylaniline acts as a precursor to triarylmethane dyes such as crystal violet and malachite green. It is also used in the synthesis of vanillin, methyl violet, and Michler's ketone. Further, it is used as a hardener for plastic resins and an acid scavenger in the manufacture of semisynthetic penicillins and cephalosporins. In addition to this, it is used as a promoter in the curing of polyester and vinyl ester resins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Dimethylaniline acts as a precursor to triarylmethane dyes such as crystal violet and malachite green. It is also used in the synthesis of vanillin, methyl violet, and Michler′s ketone. Further, it is used as a hardener for plastic resins and an acid scavenger in the manufacture of semisynthetic penicillins and cephalosporins. In addition to this, it is used as a promoter in the curing of polyester and vinyl ester resins.

Solubility
Miscible with water, ethanol, ether, acetone, chloroform, benzene and alkaline.

Notes
Incompatible with strong oxidizing agents, Strong acids, acid chlorides, acid anhydrides, chloroformates and halogens.
Specifications

Chemical Identifiers

121-69-7
121.18
JLTDJTHDQAWBAV-UHFFFAOYSA-N
949
N,N-dimethylaniline
C8H11N
MFCD00008304
dimethylaniline, dimethylphenylamine, n,n-dimethylbenzenamine, benzenamine, n,n-dimethyl, dimethylamino benzene, n,n-dimethylphenylamine, n,n-dimethylbenzeneamine, dimethylaminobenzene, dwumetyloanilina, n,n-dimethylamino benzene
CHEBI:16269
CN(C)C1=CC=CC=C1

Specifications

121-69-7
0.956
193°C
99%
C8H11N
MFCD00008304
507140
dimethylaniline, dimethylphenylamine, n,n-dimethylbenzenamine, benzenamine, n,n-dimethyl, dimethylamino benzene, n,n-dimethylphenylamine, n,n-dimethylbenzeneamine, dimethylaminobenzene, dwumetyloanilina, n,n-dimethylamino benzene
JLTDJTHDQAWBAV-UHFFFAOYSA-N
N,N-dimethylaniline
949
121.18
Liquid
2°C
7.4
62°C (143°F)
Characteristic
1.558
UN2253
14,3234
Miscible with water,ethanol,ether,acetone,chloroform,benzene and alkaline.
CN(C)C1=CC=CC=C1
121.18
CHEBI:16269
99%
N,N-Dimethylaniline
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H331-H351-H227
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Suspected of causing cancer.
Combustible liquid.

P201-P202-P210-P235-P261-P264b-P270-P271-P281-P301+P310-P302+P352-P304+P340-P308+P313-P311-P312-P330-P361-P363-P370+P378q-P501c

H227-H301+H311+H331-H351

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: N,N-DIMETHYLANILINE

missing translation for 'einecsNumber' : 204-493-5

missing translation for 'rtecsNumber' : BX4725000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Product Identifier
  • N,N-Dimethylaniline

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Carcinogenicity Category 2

Hazard Statement
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H351-Suspected of causing cancer.
  • H411-Toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

Supplemental information
  • MIXTURE LIST-Contains : N,N-Dimethylaniline

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only