Learn More
N,N-Dimethylacetamide-d{9}, 99% (Isotopic), Thermo Scientific Chemicals
248.24 € - 972.84 €
Chemical Identifiers
CAS | 116057-81-9 |
---|---|
Molecular Formula | C4H9NO |
Molecular Weight (g/mol) | 87.12 |
MDL Number | MFCD00066061 |
InChI Key | FXHOOIRPVKKKFG-UHFFFAOYSA-N |
Synonym | n,n-dimethylacetamide-d9, n,n-bis ∼2∼h_3_ methyl ∼2∼h_3_ acetamide, an,n-dimethylacetamide-d9, n,n-dimethylacetamide-d9, 99 atom % d, n,n-bis 2 h? methyl 2 h? acetamide, n,n-di 2 h? methyl 2 h? acetamide |
PubChem CID | 22789542 |
SMILES | CN(C)C(C)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11350528
|
Thermo Scientific Alfa Aesar
043854.03 |
1 g |
248.24 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11340528
|
Thermo Scientific Alfa Aesar
043854.06 |
5 g |
972.84 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
N,N-Dimethylacetamide-d{9} is a common solvent used in NMR spectroscopy.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Ampouled under argon
Chemical Identifiers
116057-81-9 | |
87.12 | |
FXHOOIRPVKKKFG-UHFFFAOYSA-N | |
22789542 |
C4H9NO | |
MFCD00066061 | |
n,n-dimethylacetamide-d9, n,n-bis ∼2∼h_3_ methyl ∼2∼h_3_ acetamide, an,n-dimethylacetamide-d9, n,n-dimethylacetamide-d9, 99 atom % d, n,n-bis 2 h? methyl 2 h? acetamide, n,n-di 2 h? methyl 2 h? acetamide | |
CN(C)C(C)=O |
Specifications
Liquid | |
1.035 g/mL | |
Ampule | |
116057-81-9 | |
165°C | |
CD3CON(CD3)2 | |
n,n-dimethylacetamide-d9, n,n-bis ∼2∼h_3_ methyl ∼2∼h_3_ acetamide, an,n-dimethylacetamide-d9, n,n-dimethylacetamide-d9, 99 atom % d, n,n-bis 2 h? methyl 2 h? acetamide, n,n-di 2 h? methyl 2 h? acetamide | |
FXHOOIRPVKKKFG-UHFFFAOYSA-N | |
2,2,2-trideuterio-N,N-bis(trideuteriomethyl)acetamide | |
22789542 | |
99% |
Colorless | |
70°C (158°F) | |
1.438 | |
-20°C | |
C4H9NO | |
MFCD00066061 | |
Miscible with water. | |
CN(C)C(C)=O | |
87.12 | |
96.19 | |
N,N-Dimethylacetamide-d9, Isotopic |
Safety and Handling
GHS H Statement
H360-H312-H332-H227
May damage fertility or the unborn child.
Harmful in contact with skin.
Harmful if inhaled.
Combustible liquid.
P201-P202-P210-P261-P271-P281-P302+P352-P304+P340-P308+P313-P312-P363-P370+P378q-P501c
H227-H312+H332-H360D
missing translation for 'tsca' : No
missing translation for 'storageNote' :
- Ampouled under argon
Recommended Storage : Ambient
RUO – Research Use Only