missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N-Dimethylacetamide, CHROMASOLV™ Plus, for HPLC, ≥99.9%, Honeywell Riedel-de Haën™
Click to view available options
Quantity:
1 L
2.5 L
4 x 2.5 L
6 x 1 L
Description
- For HPLC, spectrophotometry, and environmental analysis
- Tested for HPLC-suitability via gradient analysis and UV-VIS spectroscopy
- Low UV impurity profile
- Low impurity gradient LC baseline
- Very low levels of non-volatile impurities
Specifications
Specifications
Melting Point | -20°C (literature) |
Absorbance | 0.01 at 360 nm |
CAS Max % | 100.0000% |
Density | 0.937 g/mL at 25 °C (literature) |
Boiling Point | 164°C to 166°C |
Quantity | 6 x 1 L |
Flash Point | 64°C |
Packaging | Glass Bottle |
Linear Formula | CH3CH2COCH2CH3 |
Beilstein | 1737614 |
Show More |
Safety and Handling
Product Identifier
- N,N-Dimethylacetamide
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Reproductive toxicity Category 1B
- H312 + H332-Harmful in contact with skin or if inhaled.
- H319-Causes serious eye irritation.
- H360D-May damage the unborn child.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction