missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N-Dimethylacetamide, ≥99.5% (GC), Honeywell™ Riedel-de-Haën™
Puriss. p.a.,≥99.5% (GC)
91.31 € - 182.63 €
Chemical Identifiers
CAS | 127-19-5 |
---|---|
Molecular Formula | C4H9NO |
Molecular Weight (g/mol) | 87.12 |
MDL Number | MFCD00008686 |
InChI Key | FXHOOIRPVKKKFG-UHFFFAOYSA-N |
Synonym | dimethylacetamide, dmac, acetamide, n,n-dimethyl, acetdimethylamide, dimethyl acetamide, n,n-dimethyl acetamide, dimethylamide acetate, n,n-dimethylethanamide, dimethylacetone amide, acetyldimethylamine |
PubChem CID | 31374 |
ChEBI | CHEBI:84254 |
IUPAC Name | N,N-dimethylacetamide |
SMILES | CN(C)C(C)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15695340
|
Honeywell Riedel-de Haën
38840-1L |
1 L |
91.31 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15605350
|
Honeywell Riedel-de Haën
38840-2.5L |
2.5 L |
182.63 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
- It is the leading brand offering High-Purity Solvents for dedicated applications.
Chemical Identifiers
127-19-5 | |
87.12 | |
FXHOOIRPVKKKFG-UHFFFAOYSA-N | |
31374 | |
N,N-dimethylacetamide |
C4H9NO | |
MFCD00008686 | |
dimethylacetamide, dmac, acetamide, n,n-dimethyl, acetdimethylamide, dimethyl acetamide, n,n-dimethyl acetamide, dimethylamide acetate, n,n-dimethylethanamide, dimethylacetone amide, acetyldimethylamine | |
CHEBI:84254 | |
CN(C)C(C)=O |
Specifications
127-19-5 | |
1.8% (100°F), 11.5% (160°F) | |
914°F | |
CH3CON(CH3)2 | |
4mmHg (38°C), 2mmHg (25°C) | |
dimethylacetamide, dmac, acetamide, n,n-dimethyl, acetdimethylamide, dimethyl acetamide, n,n-dimethyl acetamide, dimethylamide acetate, n,n-dimethylethanamide, dimethylacetone amide, acetyldimethylamine | |
CN(C)C(C)=O | |
87.12 | |
CHEBI:84254 | |
≥99.5% (GC) |
0.937g/mL at 25°C | |
70°C (158°F) | |
C4H9NO | |
MFCD00008686 | |
1737614 | |
FXHOOIRPVKKKFG-UHFFFAOYSA-N | |
N,N-dimethylacetamide | |
31374 | |
87.12g/mol | |
N,N-Dimethylacetamide |
Safety and Handling
P201-P261-P280-P305 + P351 + P338-P308 + P313
missing translation for 'rtecsNumber' : 204-826-4
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- N,N-Dimethylacetamide
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Reproductive toxicity Category 1B
Hazard Statement
- H312-Harmful in contact with skin.
- H360D-May damage the unborn child.
- H332-Harmful if inhaled.
- H319-Causes serious eye irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : N,N-Dimethylacetamide
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title