missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethyl-p-phenylenediamine dihydrochloride, 99%, Thermo Scientific Chemicals

102.51 € - 294.47 €

Chemical Identifiers

CAS 536-46-9
Molecular Formula C8H14Cl2N2
Molecular Weight (g/mol) 209.11
MDL Number MFCD00012991
InChI Key PXJHVKRLFWZUNV-UHFFFAOYSA-N
Synonym n,n-dimethyl-p-phenylenediamine dihydrochloride, 4-amino-n,n-dimethylaniline dihydrochloride, n,n-dimethyl-1,4-phenylenediamine dihydrochloride, 1,4-benzenediamine, n,n-dimethyl-, dihydrochloride, 1,4-amino-n,n-dimethylaniline,dihydrochloride, n,n-dimethyl-1,4-benzenediamine dihydrochloride, n,n-dimethyl-p-phenylenediamine 2hcl, dimethyl-p-phenylenediamine dihydrochloride, n,n-dimethyl-p-phenylenediammonium dichloride, pubchem21651
PubChem CID 2724166
IUPAC Name 4-N,4-N-dimethylbenzene-1,4-diamine;dihydrochloride
SMILES [H+].[H+].[Cl-].[Cl-].CNC1=CC=C(NC)C=C1
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10378050
View Documents
Thermo Scientific
204350250
25 g Glass bottle
102.51 €
25g
Please to purchase this item. Need a web account? Register with us today!
10043573
View Documents
Thermo Scientific Acros
204351000
100 g Glass bottle
294.47 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

536-46-9
209.11
PXJHVKRLFWZUNV-UHFFFAOYSA-N
2724166
[H+].[H+].[Cl-].[Cl-].CNC1=CC=C(NC)C=C1
C8H14Cl2N2
MFCD00012991
n,n-dimethyl-p-phenylenediamine dihydrochloride, 4-amino-n,n-dimethylaniline dihydrochloride, n,n-dimethyl-1,4-phenylenediamine dihydrochloride, 1,4-benzenediamine, n,n-dimethyl-, dihydrochloride, 1,4-amino-n,n-dimethylaniline,dihydrochloride, n,n-dimethyl-1,4-benzenediamine dihydrochloride, n,n-dimethyl-p-phenylenediamine 2hcl, dimethyl-p-phenylenediamine dihydrochloride, n,n-dimethyl-p-phenylenediammonium dichloride, pubchem21651
4-N,4-N-dimethylbenzene-1,4-diamine;dihydrochloride

Specifications

536-46-9
100.0
Authentic
C8H14Cl2N2
MFCD00012991
01,293
n,n-dimethyl-p-phenylenediamine dihydrochloride, 4-amino-n,n-dimethylaniline dihydrochloride, n,n-dimethyl-1,4-phenylenediamine dihydrochloride, 1,4-benzenediamine, n,n-dimethyl-, dihydrochloride, 1,4-amino-n,n-dimethylaniline,dihydrochloride, n,n-dimethyl-1,4-benzenediamine dihydrochloride, n,n-dimethyl-p-phenylenediamine 2hcl, dimethyl-p-phenylenediamine dihydrochloride, n,n-dimethyl-p-phenylenediammonium dichloride, pubchem21651
PXJHVKRLFWZUNV-UHFFFAOYSA-N
4-N,4-N-dimethylbenzene-1,4-diamine;dihydrochloride
2724166
99%
N,N-Dimethyl-p-phenylenediamine dihydrochloride
98.5
222.0°C
99%
(CH3)2NC6H4NH2·2HCl
13, 73
10, 3248
(5% in water) Clear colorless to purple-brown
[H+].[H+].[Cl-].[Cl-].CNC1=CC=C(NC)C=C1
209.11
209.12
Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Fatal if swallowed.
Toxic in contact with skin.
Fatal if inhaled.
Causes serious eye irritation.

GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 208-635-7

Product Identifier
  • N,N-Dimethyl-p-phenylenediamine dihydrochloride

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H319-Causes serious eye irritation.
  • H330-Fatal if inhaled.
  • H311-Toxic in contact with skin.
  • H300-Fatal if swallowed.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only