missing translation for 'onlineSavingsMsg'
Learn More
N,N-Dimethyl-L-phenylalanine, 99%, Thermo Scientific Chemicals Product Code.: 11985361

N,N-Dimethyl-L-phenylalanine, 99%, Thermo Scientific Chemicals

Product Code. 11985361
1 g, Glass bottle
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 11985361

Brand: Thermo Scientific Acros 440030010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17469-89-5
Molecular Formula C11H15NO2
Molecular Weight (g/mol) 193.25
MDL Number MFCD00082539
InChI Key HOGIQTACRLIOHC-JTQLQIEISA-N
Synonym n,n-dimethyl-l-phenylalanine, phenylalanine, n,n-dimethyl, n,n-dimethylphenylalanine, l-phenylalanine, n,n-dimethyl, s-2-dimethylamino-3-phenylpropanoic acid, 2s-2-dimethylamino-3-phenylpropanoic acid, alanine, n,n-dimethyl-3-phenyl-, l, l-phenylalanine,n,n-dimethyl, s-2-dimethylamino-3-phenyl-propionic acidShow MoreShow Less
PubChem CID 11830289
IUPAC Name (2S)-2-(dimethylamino)-3-phenylpropanoic acid
SMILES CN(C)[C@@H](CC1=CC=CC=C1)C(O)=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 225.0°C to 227.0°C
Color White
Density 0.0950g/mL
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Specific Gravity 0.095
Specific Rotation 73.92
Solubility Information Solubility in water: in water:(20°C) freely soluble
Formula Weight 193.25
Percent Purity 99%
Physical Form Powder
Chemical Name or Material N, N-Dimethyl-L-phenylalanine
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number345340 Certificate TypeCertificate of Analysis Date17/04/2024 Catalog Number440030050,440030010
Lot Number345114 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number440030050,440030010
Lot Number344352 Certificate TypeCertificate of Analysis Date09/01/2023 Catalog Number440030050,440030010
Lot Number343715 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number440030050,440030010
Lot Number344022 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number440030050,440030010

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
N,N-Dimethyl-L-phenylalanine, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.