missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethyl-3-nitroaniline, 98%, Thermo Scientific Chemicals

26.00 € - 77.00 €

Chemical Identifiers

CAS 619-31-8
Molecular Formula C8H10N2O2
Molecular Weight (g/mol) 166.18
MDL Number MFCD00007236
InChI Key CJDICMLSLYHRPT-UHFFFAOYSA-N
Synonym 3-nitro-n,n-dimethylaniline, benzenamine, n,n-dimethyl-3-nitro, m-nitro-n,n-dimethylaniline, n,n-dimethyl-m-nitroaniline, 1-dimethylamino-3-nitrobenzene, benzene, 1-dimethylamino-3-nitro, aniline, n,n-dimethyl-m-nitro, dimethyl-3-nitro-phenyl-amine, dimethyl 3-nitrophenyl amine, m-nitrodimethylaniline
PubChem CID 69269
IUPAC Name N,N-dimethyl-3-nitroaniline
SMILES CN(C)C1=CC(=CC=C1)[N+](=O)[O-]
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10550571
View Documents
Thermo Scientific Acros
116470050
5 g Glass bottle
26.00 €
5g
Please to purchase this item. Need a web account? Register with us today!
10656492
View Documents
Thermo Scientific Acros
116470250
25 g Glass bottle
77.00 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

619-31-8
166.18
CJDICMLSLYHRPT-UHFFFAOYSA-N
69269
CN(C)C1=CC(=CC=C1)[N+](=O)[O-]
C8H10N2O2
MFCD00007236
3-nitro-n,n-dimethylaniline, benzenamine, n,n-dimethyl-3-nitro, m-nitro-n,n-dimethylaniline, n,n-dimethyl-m-nitroaniline, 1-dimethylamino-3-nitrobenzene, benzene, 1-dimethylamino-3-nitro, aniline, n,n-dimethyl-m-nitro, dimethyl-3-nitro-phenyl-amine, dimethyl 3-nitrophenyl amine, m-nitrodimethylaniline
N,N-dimethyl-3-nitroaniline

Specifications

619-31-8
100.0
Orange to Orange-Red
98%
O2NC6H4N(CH3)2
12, 701
Solubility in water: insoluble
CN(C)C1=CC(=CC=C1)[N+](=O)[O-]
166.18
166.18
Crystalline Powder or Crystals
97.5
57°C to 61°C
Authentic
C8H10N2O2
MFCD00007236
3-nitro-n,n-dimethylaniline, benzenamine, n,n-dimethyl-3-nitro, m-nitro-n,n-dimethylaniline, n,n-dimethyl-m-nitroaniline, 1-dimethylamino-3-nitrobenzene, benzene, 1-dimethylamino-3-nitro, aniline, n,n-dimethyl-m-nitro, dimethyl-3-nitro-phenyl-amine, dimethyl 3-nitrophenyl amine, m-nitrodimethylaniline
CJDICMLSLYHRPT-UHFFFAOYSA-N
N,N-dimethyl-3-nitroaniline
69269
98%
N, N-Dimethyl-3-nitroaniline, 98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 210-590-3

missing translation for 'tsca' : TSCA

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only