Learn More
N,N-Dimethyl-1-naphthylamine, 99%, Thermo Scientific Chemicals
32.30 € - 110.00 €
Chemical Identifiers
CAS | 86-56-6 |
---|---|
Molecular Formula | C12H13N |
Molecular Weight (g/mol) | 171.243 |
MDL Number | MFCD00003919 |
InChI Key | AJUXDFHPVZQOGF-UHFFFAOYSA-N |
Synonym | n,n-dimethyl-1-naphthylamine, 1-dimethylaminonaphthalene, 1-naphthalenamine, n,n-dimethyl, n,n-dimethyl-1-naphthalenamine, n,n-dimethyl-1-napthylamine, dimethyl 1-naphthyl amine, alpha-dimethylaminonaphthalene, 1-naphthylamine, n,n-dimethyl, dimethyl-alpha-naphthylamine, n,n-dimethyl-1-naftylamin |
PubChem CID | 6848 |
IUPAC Name | N,N-dimethylnaphthalen-1-amine |
SMILES | CN(C)C1=CC=CC2=CC=CC=C21 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11337908
|
Thermo Scientific Alfa Aesar
036438.AA |
5 mL |
32.30 €
5mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11347908
|
Thermo Scientific Alfa Aesar
036438.AC |
25 mL |
110.00 €
25mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
N,N-Dimethyl-1-naphthylamine is used in the nitrate reductase test to form a red precipitate of Prontosil by reacting with a nitrite-sulfanilic acid complex. And is also used to detection and determination of nitrites.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
86-56-6 | |
171.243 | |
AJUXDFHPVZQOGF-UHFFFAOYSA-N | |
6848 | |
CN(C)C1=CC=CC2=CC=CC=C21 |
C12H13N | |
MFCD00003919 | |
n,n-dimethyl-1-naphthylamine, 1-dimethylaminonaphthalene, 1-naphthalenamine, n,n-dimethyl, n,n-dimethyl-1-naphthalenamine, n,n-dimethyl-1-napthylamine, dimethyl 1-naphthyl amine, alpha-dimethylaminonaphthalene, 1-naphthylamine, n,n-dimethyl, dimethyl-alpha-naphthylamine, n,n-dimethyl-1-naftylamin | |
N,N-dimethylnaphthalen-1-amine |
Specifications
86-56-6 | |
1.039 g/mL | |
C12H13N | |
MFCD00003919 | |
14,3252 | |
Insoluble in water. | |
CN(C)C1=CC=CC2=CC=CC=C21 | |
171.243 | |
171.24 | |
Liquid |
Yellow | |
145°C to 147°C (12 mmHg) | |
1.623 | |
UN3082 | |
n,n-dimethyl-1-naphthylamine, 1-dimethylaminonaphthalene, 1-naphthalenamine, n,n-dimethyl, n,n-dimethyl-1-naphthalenamine, n,n-dimethyl-1-napthylamine, dimethyl 1-naphthyl amine, alpha-dimethylaminonaphthalene, 1-naphthylamine, n,n-dimethyl, dimethyl-alpha-naphthylamine, n,n-dimethyl-1-naftylamin | |
AJUXDFHPVZQOGF-UHFFFAOYSA-N | |
N,N-dimethylnaphthalen-1-amine | |
6848 | |
99% | |
N,N-Dimethyl-1-naphthylamine |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
missing translation for 'einecsNumber' : 201-682-4
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only