missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diethyl-p-phenylenediamine, 98%, Thermo Scientific Chemicals

Product Code. 10435675
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93-05-0
MDL Number MFCD00007861
InChI Key QNGVNLMMEQUVQK-UHFFFAOYSA-N
Synonym n,n-diethyl-1,4-phenylenediamine, 4-amino-n,n-diethylaniline, n,n-diethyl-p-phenylenediamine, n1,n1-diethylbenzene-1,4-diamine, p-aminodiethylaniline, 4-diethylamino aniline, p-amino-n,n-diethylaniline, n,n-diethyl-1,4-benzenediamine, diethylaminoaniline, p-diethylamino aniline
PubChem CID 7120
IUPAC Name 4-N,4-N-diethylbenzene-1,4-diamine
SMILES CCN(CC)C1=CC=C(C=C1)N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 23.0°C
Color Brown to Brown-Yellow
Density 0.9880g/mL
Boiling Point 268.0°C
Flash Point 139°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C10H16N2
Linear Formula (CH3CH2)2NC6H4NH2
Refractive Index 1.5690 to 1.5730 (20°C, 589nm)
Quantity 100 g
Molecular Weight (g/mol) 164.252
Formula Weight 164.25
Percent Purity 98%
Physical Form Liquid or Low Melting Solid
Chemical Name or Material N, N-Diethyl-p-phenylenediamine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • N,N-Diethyl-p-phenylenediamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'