Learn More
N,N-Diethyl(2-chloro-1,1,2-trifluoroethyl)amine, 90%, Thermo Scientific Chemicals
41.60 € - 373.00 €
Chemical Identifiers
CAS | 357-83-5 |
---|---|
Molecular Formula | C6H11ClF3N |
Molecular Weight (g/mol) | 189.606 |
MDL Number | MFCD00054672 |
InChI Key | BDZHKUAKSMWSAJ-UHFFFAOYSA-N |
Synonym | 2-chloro-1,1,2-trifluoroethyl diethylamine, n,n-diethyl 2-chloro-1,1,2-trifluoroethyl amine, n-2-chloro-1,1,2-trifluoroethyl diethylamine, yarovenko's reagent, acmc-20appo, 2-chloro-1,1,2-trifluorotriethylamine, 1,1,2-trifluoro-2-chloroethyldiethylamine, 2-chloro-1,1,2-trifluoroethyldiethylamine, ethanamine, 2-chloro-n,n-diethyltrifluoro, 1-diethylamino-1,1,2trifluoro-2-chloroethane |
PubChem CID | 136184 |
IUPAC Name | 2-chloro-N,N-diethyl-1,1,2-trifluoroethanamine |
SMILES | CCN(CC)C(C(F)Cl)(F)F |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11338386
|
Thermo Scientific Alfa Aesar
L17330.06 |
5 g |
41.60 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11348386
|
Thermo Scientific Alfa Aesar
L17330.14 |
25 g |
113.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11358386
|
Thermo Scientific Alfa Aesar
L17330.22 |
100 g |
373.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
357-83-5 | |
189.606 | |
BDZHKUAKSMWSAJ-UHFFFAOYSA-N | |
136184 | |
CCN(CC)C(C(F)Cl)(F)F |
C6H11ClF3N | |
MFCD00054672 | |
2-chloro-1,1,2-trifluoroethyl diethylamine, n,n-diethyl 2-chloro-1,1,2-trifluoroethyl amine, n-2-chloro-1,1,2-trifluoroethyl diethylamine, yarovenko's reagent, acmc-20appo, 2-chloro-1,1,2-trifluorotriethylamine, 1,1,2-trifluoro-2-chloroethyldiethylamine, 2-chloro-1,1,2-trifluoroethyldiethylamine, ethanamine, 2-chloro-n,n-diethyltrifluoro, 1-diethylamino-1,1,2trifluoro-2-chloroethane | |
2-chloro-N,N-diethyl-1,1,2-trifluoroethanamine |
Specifications
357-83-5 | |
38°C (10 mmHg) | |
ClFCHCF2N(CH2CH3)2 | |
UN3267 | |
Moisture sensitive | |
BDZHKUAKSMWSAJ-UHFFFAOYSA-N | |
2-chloro-N,N-diethyl-1,1,2-trifluoroethanamine | |
136184 | |
90% |
1.19 | |
C6H11ClF3N | |
MFCD00054672 | |
1098675 | |
2-chloro-1,1,2-trifluoroethyl diethylamine, n,n-diethyl 2-chloro-1,1,2-trifluoroethyl amine, n-2-chloro-1,1,2-trifluoroethyl diethylamine, yarovenko's reagent, acmc-20appo, 2-chloro-1,1,2-trifluorotriethylamine, 1,1,2-trifluoro-2-chloroethyldiethylamine, 2-chloro-1,1,2-trifluoroethyldiethylamine, ethanamine, 2-chloro-n,n-diethyltrifluoro, 1-diethylamino-1,1,2trifluoro-2-chloroethane | |
CCN(CC)C(C(F)Cl)(F)F | |
189.606 | |
189.61 | |
N,N-Diethyl(2-chloro-1,1,2-trifluoroethyl)amine |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314
missing translation for 'dotInformation' : Hazard Class: 8; Packaging Group: III
missing translation for 'einecsNumber' : 000-000-0
missing translation for 'tsca' : No
Recommended Storage : Keep cold
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.