missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dibenzyl-4-bromoaniline, 95%, Thermo Scientific™

Product Code. 11382269
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65145-14-4
Molecular Formula C20H18BrN
Molecular Weight (g/mol) 352.28
MDL Number MFCD09037473
InChI Key RSFLPIGKWXARAL-UHFFFAOYSA-N
Synonym 4-bromo-n,n-dibenzylaniline, benzenemethanamine, n-4-bromophenyl-n-phenylmethyl, acmc-20ak9j, 4-bromophenyldibenzylamine, n-4-bromophenyl dibenzylamine, n,n-dibenzyl-4-bromobenzenamine, 4-n,n-dibenzylamino bromobenzene, n-4bromophenyl-n-phenylmethyl benzenemethanamine, benzenemethanamine,n-4-bromophenyl-n-phenylmethyl
PubChem CID 285423
IUPAC Name N,N-dibenzyl-4-bromoaniline
SMILES BrC1=CC=C(C=C1)N(CC1=CC=CC=C1)CC1=CC=CC=C1

Specifications

Quantity 1 g
Formula Weight 352.28
Percent Purity 95%
Chemical Name or Material N,N-Dibenzyl-4-bromoaniline
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'