missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Di(1-naphthyl)-N,N'-diphenylbenzidine, 98%, Thermo Scientific Chemicals

Product Code. 15421749
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123847-85-8
Molecular Formula C44H32N2
Molecular Weight (g/mol) 588.754
MDL Number MFCD03093246
InChI Key IBHBKWKFFTZAHE-UHFFFAOYSA-N
Synonym n4,n4'-di naphthalen-1-yl-n4,n4'-diphenyl-1,1'-biphenyl-4,4'-diamine, alpha-npb, n,n'-bis naphthalen-1-yl-n,n'-bis phenyl benzidine, 4,4'-bis n-1-naphthyl-n-phenylamino biphenyl, n,n'-di 1-naphthyl-n,n'-diphenylbenzidine, n,n'-bis-1-naphthalenyl-n,n'-bis-phenyl-1,1'-biphenyl-4,4'-diamine, 1,1'-biphenyl-4,4'-diamine, n,n'-di-1-naphthalenyl-n,n'-diphenyl, n,n'-di-1-naphthyl-n,n'-diphenylbenzidine
PubChem CID 5069127
IUPAC Name N-[4-[4-(N-naphthalen-1-ylanilino)phenyl]phenyl]-N-phenylnaphthalen-1-amine
SMILES C1=CC=C(C=C1)N(C2=CC=C(C=C2)C3=CC=C(C=C3)N(C4=CC=CC=C4)C5=CC=CC6=CC=CC=C65)C7=CC=CC8=CC=CC=C87

Specifications

Melting Point 279°C to 283°C
Color Yellow
Quantity 5 g
Beilstein 8022378
Formula Weight 588.75
Percent Purity 98%
Physical Form Powder
Chemical Name or Material N,N'-Di(1-naphthyl)-N,N'-diphenylbenzidine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'