missing translation for 'onlineSavingsMsg'
Learn More

N-(n-Butyl)ethylenediamine, 97%, Thermo Scientific Chemicals

92.80 € - 358.00 €

Chemical Identifiers

CAS 19522-69-1
Molecular Formula C6H16N2
Molecular Weight (g/mol) 116.208
MDL Number MFCD00025602
InChI Key DFPGBRPWDZFIPP-UHFFFAOYSA-N
Synonym n-butylethylenediamine, 2-butylamino ethylamine, n1-butylethane-1,2-diamine, 2-aminoethyl butyl amine, 1,2-ethanediamine,n1-butyl, 2-butylamino-ethylamine, 2-n-butylaminoethylamine, acmc-1bs5m, n-butyl-1,2-ethanediamine, n-butylethane-1,2-diamine
PubChem CID 519668
IUPAC Name N'-butylethane-1,2-diamine
SMILES CCCCNCCN
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11303067
View Documents
Thermo Scientific Alfa Aesar
L13437.06
5 g
92.80 €
5g
Please to purchase this item. Need a web account? Register with us today!
11313067
View Documents
Thermo Scientific Alfa Aesar
L13437.14
25 g
358.00 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

19522-69-1
116.208
DFPGBRPWDZFIPP-UHFFFAOYSA-N
519668
CCCCNCCN
C6H16N2
MFCD00025602
n-butylethylenediamine, 2-butylamino ethylamine, n1-butylethane-1,2-diamine, 2-aminoethyl butyl amine, 1,2-ethanediamine,n1-butyl, 2-butylamino-ethylamine, 2-n-butylaminoethylamine, acmc-1bs5m, n-butyl-1,2-ethanediamine, n-butylethane-1,2-diamine
N'-butylethane-1,2-diamine

Specifications

19522-69-1
170°C to 171°C
C6H16N2
MFCD00025602
1732700
n-butylethylenediamine, 2-butylamino ethylamine, n1-butylethane-1,2-diamine, 2-aminoethyl butyl amine, 1,2-ethanediamine,n1-butyl, 2-butylamino-ethylamine, 2-n-butylaminoethylamine, acmc-1bs5m, n-butyl-1,2-ethanediamine, n-butylethane-1,2-diamine
CCCCNCCN
116.208
116.21
N-(n-Butyl)ethylenediamine
0.836
59°C (138°F)
1.443
UN2734
Air Sensitive
DFPGBRPWDZFIPP-UHFFFAOYSA-N
N'-butylethane-1,2-diamine
519668
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H226-H314-H335

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, FLAMMABLE, N.O.S.

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.