missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-2-phenoxyethylamine, 98%, Thermo Scientific Chemicals

Product Code. 11459390
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37421-04-8
Molecular Formula C9H13NO
Molecular Weight (g/mol) 151.21
MDL Number MFCD00085690
InChI Key GUMBNOITRNRLCS-UHFFFAOYSA-N
Synonym methyl 2-phenoxyethyl amine, n-methyl-2-phenoxyethylamine, methyl-2-phenoxy-ethyl-amine, n-methyl-2-phenoxy-ethylamin, ethylamine, n-methyl-2-phenoxy, ethanamine, n-methyl-2-phenoxy, n-methyl-n-2-phenoxyethyl amine, alpha-phenoxy-beta-methylaminoethane, acmc-1ae2y, n-methyl-2-phenoxyethylamin
PubChem CID 37732
IUPAC Name N-methyl-2-phenoxyethanamine
SMILES CNCCOC1=CC=CC=C1

Specifications

Linear Formula C6H5OCH2CH2NHCH3
Quantity 250 mg
UN Number UN2735
Formula Weight 151.21
Percent Purity 98%
Chemical Name or Material N-Methyl-2-phenoxyethylamine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'