missing translation for 'onlineSavingsMsg'
Learn More

N-Isopropylmethylamine, 98%, Thermo Scientific Chemicals

85.20 € - 316.00 €

Chemical Identifiers

CAS 4747-21-1
Molecular Formula C4H11N
Molecular Weight (g/mol) 73.139
MDL Number MFCD00042859
InChI Key XHFGWHUWQXTGAT-UHFFFAOYSA-N
Synonym n-isopropylmethylamine, n-methylisopropylamine, methylisopropylamine, 2-propanamine, n-methyl, isopropylmethylamine, 2-methylaminopropane, methyl propan-2-yl amine, ethylamine, n,1-dimethyl, unii-j3ol90426g, n-methylisopropyylamine
PubChem CID 78485
IUPAC Name N-methylpropan-2-amine
SMILES CC(C)NC
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15442659
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H65892.06
5 g
85.20 €
5g
Please to purchase this item. Need a web account? Register with us today!
15452659
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H65892.14
25 g
316.00 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

N-Isopropylmethylamine is used in the generation of in situ micelles having distinct sizes, by sequential self-assembly of a series of stimuli-responsive amphiphilic block copolymers, prepared by reversible addition-fragmentation chain transfer (RAFT) polymerization. It is suitable for use in the synthesis of novel 5-ethoxy-N,N-dialkyl-[α,α,β,β-H4]-tryptamines and their [α,α,β,β,β-D4]-counterparts following the Speeter and Anthony procedure.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Isopropylmethylamine is used in the generation of in situ micelles having distinct sizes, by sequential self-assembly of a series of stimuli-responsive amphiphilic block copolymers, prepared by reversible addition-fragmentation chain transfer (RAFT) polymerization. It is suitable for use in the synthesis of novel 5-ethoxy-N,N-dialkyl-[α,α,β,β-H4]-tryptamines and their [α,α,β,β,β-D4]-counterparts following the Speeter and Anthony procedure.

Solubility
Soluble in chloroform, DMSO, methanol.

Notes
Air Sensitive. Store away from air. Store under dry inert gas. Do not store together with acids. Incompatible with acids , air and oxidizing agents.
Specifications

Chemical Identifiers

4747-21-1
73.139
XHFGWHUWQXTGAT-UHFFFAOYSA-N
78485
CC(C)NC
C4H11N
MFCD00042859
n-isopropylmethylamine, n-methylisopropylamine, methylisopropylamine, 2-propanamine, n-methyl, isopropylmethylamine, 2-methylaminopropane, methyl propan-2-yl amine, ethylamine, n,1-dimethyl, unii-j3ol90426g, n-methylisopropyylamine
N-methylpropan-2-amine

Specifications

4747-21-1
0.702 g/mL
-32°C (-26°F)
(CH3)2CHNHCH3
MFCD00042859
1730877
n-isopropylmethylamine, n-methylisopropylamine, methylisopropylamine, 2-propanamine, n-methyl, isopropylmethylamine, 2-methylaminopropane, methyl propan-2-yl amine, ethylamine, n,1-dimethyl, unii-j3ol90426g, n-methylisopropyylamine
XHFGWHUWQXTGAT-UHFFFAOYSA-N
N-methylpropan-2-amine
78485
98%
N-Isopropylmethylamine
Colorless
50°C to 53°C
C4H11N
1.348
UN2734
Air Sensitive
Soluble in chloroform,DMSO,methanol.
CC(C)NC
73.139
73.14
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c

H225-H301+H331-H314-H335

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packaging Group: I; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, FLAMMABLE, N.O.S.

missing translation for 'einecsNumber' : 225-266-7

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures; Store under Argon

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only