Learn More
N-Isopropylaniline, 98%, Thermo Scientific Chemicals
43.70 € - 137.00 €
Chemical Identifiers
CAS | 768-52-5 |
---|---|
Molecular Formula | C9H13N |
Molecular Weight (g/mol) | 135.21 |
MDL Number | MFCD00026347 |
InChI Key | FRCFWPVMFJMNDP-UHFFFAOYSA-N |
Synonym | n-isopropylaniline, isopropylaniline, n-phenylisopropylamine, benzenamine, n-1-methylethyl, aniline, n-isopropyl, n-propan-2-yl aniline, n-iso-propylaniline, unii-i7y9616f3r, ccris 4831, 2-phenylamino propane |
PubChem CID | 13032 |
IUPAC Name | N-propan-2-ylaniline |
SMILES | CC(C)NC1=CC=CC=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11322926
|
Thermo Scientific Alfa Aesar
L10646.09 |
10 g |
43.70 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11332926
|
Thermo Scientific Alfa Aesar
L10646.18 |
50 g |
137.00 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
768-52-5 | |
135.21 | |
FRCFWPVMFJMNDP-UHFFFAOYSA-N | |
13032 | |
CC(C)NC1=CC=CC=C1 |
C9H13N | |
MFCD00026347 | |
n-isopropylaniline, isopropylaniline, n-phenylisopropylamine, benzenamine, n-1-methylethyl, aniline, n-isopropyl, n-propan-2-yl aniline, n-iso-propylaniline, unii-i7y9616f3r, ccris 4831, 2-phenylamino propane | |
N-propan-2-ylaniline |
Specifications
768-52-5 | |
0.938 | |
88°C (190°F) | |
1.5385 | |
UN2810 | |
n-isopropylaniline, isopropylaniline, n-phenylisopropylamine, benzenamine, n-1-methylethyl, aniline, n-isopropyl, n-propan-2-yl aniline, n-iso-propylaniline, unii-i7y9616f3r, ccris 4831, 2-phenylamino propane | |
CC(C)NC1=CC=CC=C1 | |
135.21 | |
135.21 | |
N-Isopropylaniline |
-32°C | |
203°C to 204°C | |
C9H13N | |
MFCD00026347 | |
2205871 | |
FRCFWPVMFJMNDP-UHFFFAOYSA-N | |
N-propan-2-ylaniline | |
13032 | |
98% |
Safety and Handling
GHS H Statement
H331-H227-H302
Toxic if inhaled.
Combustible liquid.
Harmful if swallowed.
P210-P235-P260-P264b-P270-P271-P280-P284-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P330-P332+P313-P362-P370+P378q-P501c
H227-H302-H315-H319-H330
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 212-196-7
missing translation for 'rtecsNumber' : BY4190000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.