missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Heptane, Extra Pure, SLR, Fisher Chemical™
32.80 € - 635.00 €
Chemical Identifiers
CAS | 142-82-5 |
---|---|
Molecular Formula | C7H16 |
Molecular Weight (g/mol) | 100.21 |
MDL Number | MFCD00009544 |
InChI Key | IMNFDUFMRHMDMM-UHFFFAOYSA-N |
Synonym | n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish |
PubChem CID | 8900 |
ChEBI | CHEBI:43098 |
IUPAC Name | heptane |
SMILES | CCCCCCC |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10417450
|
Fisher Chemical
H/0155/08 |
500 mL | Amber Glass Bottle |
32.80 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12606717
|
Fisher Chemical
H/0155/15 |
1 L | Amber Glass Bottle |
36.50 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10234530
|
Fisher Chemical
H/0155/17 |
2.5 L | Amber Glass Bottle |
110.00 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12616717
|
Fisher Chemical
H/0155/PB17 |
2.5 L | Plastic-coated Glass Bottle |
108.00 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10417640
|
Fisher Chemical
H/0155/21 |
5 L | HDPE Bottle |
179.00 €
5 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10232790
|
Fisher Chemical
H/0155/FP21 |
5 L | Plastic Container |
129.00 €
5 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10365750
|
Fisher Chemical
H/0155/25 |
25 L | Metal Can |
635.00 €
25 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
142-82-5 | |
100.21 | |
IMNFDUFMRHMDMM-UHFFFAOYSA-N | |
8900 | |
heptane |
C7H16 | |
MFCD00009544 | |
n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish | |
CHEBI:43098 | |
CCCCCCC |
Specifications
-91°C | |
Colorless | |
Liquid | |
MFCD00009544 | |
0.4 mPaS at 20°C | |
IMNFDUFMRHMDMM-UHFFFAOYSA-N | |
heptane | |
8900 | |
100.2g/mol |
142-82-5 | |
98°C | |
C7H16 | |
48mbar at 20°C | |
n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish | |
CCCCCCC | |
100.21 | |
CHEBI:43098 |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- n-Heptane
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- Aspiration toxicity Category 1
- LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H315-Causes skin irritation.
- H336-May cause drowsiness or dizziness.
- H304-May be fatal if swallowed and enters airways.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P331-Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P273-Avoid release to the environment.