missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Fmoc-3-(4-pyridyl)-L-alanine, 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
Fmoc-(4-pyridyl)-Ala-OH is potentially useful for solid phase peptide synthesis techniques.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Fmoc-∫-(4-pyridyl)-Ala-OH is potentially useful for solid phase peptide synthesis techniques.
Solubility
Soluble in 0.1g in 4mL DMF(clearly soluble).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents. Store at 4°C.
Fmoc-∫-(4-pyridyl)-Ala-OH is potentially useful for solid phase peptide synthesis techniques.
Solubility
Soluble in 0.1g in 4mL DMF(clearly soluble).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents. Store at 4°C.
Specifications
Specifications
Quantity | 5 g |
Beilstein | 7316905 |
Solubility Information | Soluble in 0.1g in 4mL DMF(clearly soluble). |
Formula Weight | 388.42 |
Percent Purity | 95% |
Chemical Name or Material | N-Fmoc-3-(4-pyridyl)-L-alanine |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'