Learn More
N-Ethylethylenediamine, 98%, Thermo Scientific Chemicals
Description
N-Ethylethylenediamine is a chelating agent used in mutagenesis research studies. It undergoes condensation with 2-hydroxy-3-methoxybenzaldehyde. It also forms organometallic ruthenium(II) complexes that inhibit the growth in human ovarian cancer cell lines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
N-Ethylethylenediamine is a chelating agent used in mutagenesis research studies. It undergoes condensation with 2-hydroxy-3-methoxybenzaldehyde. It also forms organometallic ruthenium(II) complexes that inhibit the growth in human ovarian cancer cell lines.
Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents.
Specifications
Specifications
Density | 0.837 |
Boiling Point | 128°C to 130°C |
Flash Point | 34°C (93°F) |
Refractive Index | 1.44 |
Quantity | 25 g |
UN Number | UN2733 |
Beilstein | 1098272 |
Sensitivity | Air Sensitive |
Formula Weight | 88.15 |
Percent Purity | 98% |
Show More |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.