missing translation for 'onlineSavingsMsg'
Learn More

n-Butyltin trichloride, 96%, Thermo Scientific Chemicals

37.80 € - 96.70 €

Chemical Identifiers

CAS 1118-46-3
Molecular Formula C4H9Cl3Sn
Molecular Weight (g/mol) 282.176
MDL Number MFCD00000515
InChI Key YMLFYGFCXGNERH-UHFFFAOYSA-K
Synonym butyltin trichloride, stannane, butyltrichloro, monobutyltin trichloride, butyltrichlorotin, trichlorobutyltin, butyl trichloro stannane, n-butyltin trichloride, trichlorobutylstannane, stannane, trichlorobutyl, monotributyltin trichloride
PubChem CID 14236
IUPAC Name butyl(trichloro)stannane
SMILES CCCC[Sn](Cl)(Cl)Cl
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11314819
View Documents Promotion Details
Thermo Scientific Alfa Aesar
071125.14
25 g
37.80 €
25g
Please to purchase this item. Need a web account? Register with us today!
11304819
View Documents Promotion Details
Thermo Scientific Alfa Aesar
071125.22
100 g
96.70 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1118-46-3
282.176
YMLFYGFCXGNERH-UHFFFAOYSA-K
14236
CCCC[Sn](Cl)(Cl)Cl
C4H9Cl3Sn
MFCD00000515
butyltin trichloride, stannane, butyltrichloro, monobutyltin trichloride, butyltrichlorotin, trichlorobutyltin, butyl trichloro stannane, n-butyltin trichloride, trichlorobutylstannane, stannane, trichlorobutyl, monotributyltin trichloride
butyl(trichloro)stannane

Specifications

1118-46-3
1.693 g/mL
81°C (178°F)
C4H9Cl3Sn
MFCD00000515
Moisture sensitive
YMLFYGFCXGNERH-UHFFFAOYSA-K
butyl(trichloro)stannane
14236
96%
n-Butyltin trichloride
-63°C
98°C (10 mmHg)
Acrid
1.5244
UN3265
butyltin trichloride, stannane, butyltrichloro, monobutyltin trichloride, butyltrichlorotin, trichlorobutyltin, butyl trichloro stannane, n-butyltin trichloride, trichlorobutylstannane, stannane, trichlorobutyl, monotributyltin trichloride
CCCC[Sn](Cl)(Cl)Cl
282.176
282.17
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H372-H314-H318-H312-H227
Causes damage to organs through prolonged or repeated exposure.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful in contact with skin.
Combustible liquid.

P210-P235-P260-P264b-P271-P280-P284-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H227-H314

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

missing translation for 'einecsNumber' : 214-263-6

missing translation for 'rtecsNumber' : WH6780000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only