missing translation for 'onlineSavingsMsg'
Learn More
Learn More
n-Butyl heptafluorobutyrate, 97%, Thermo Scientific™
19.00 € - 68.00 €
Chemical Identifiers
CAS | 1559-07-5 |
---|---|
Molecular Formula | C8H9F7O2 |
Molecular Weight (g/mol) | 270.147 |
MDL Number | MFCD00039240 |
InChI Key | YDXXJZKOOOJVEE-UHFFFAOYSA-N |
Synonym | n-butyl heptafluorobutyrate, butyric acid, heptafluoro-, butyl ester, butyl heptafluorobutanoate, heptafluorobutyric acid, n-butyl ester, n-butylheptafluorobutyrate, butyl perfluorobutanoat, butyl perfluorobutyrate, butylheptafluorobutyrate, butyl heptafluorobutyrate, n-butyl perfluoro-n-butyrate |
PubChem CID | 137079 |
IUPAC Name | butyl 2,2,3,3,4,4,4-heptafluorobutanoate |
SMILES | CCCCOC(=O)C(C(C(F)(F)F)(F)F)(F)F |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
1559-07-5 | |
270.147 | |
YDXXJZKOOOJVEE-UHFFFAOYSA-N | |
137079 | |
CCCCOC(=O)C(C(C(F)(F)F)(F)F)(F)F |
C8H9F7O2 | |
MFCD00039240 | |
n-butyl heptafluorobutyrate, butyric acid, heptafluoro-, butyl ester, butyl heptafluorobutanoate, heptafluorobutyric acid, n-butyl ester, n-butylheptafluorobutyrate, butyl perfluorobutanoat, butyl perfluorobutyrate, butylheptafluorobutyrate, butyl heptafluorobutyrate, n-butyl perfluoro-n-butyrate | |
butyl 2,2,3,3,4,4,4-heptafluorobutanoate |
Specifications
1559-07-5 | |
131°C to 133°C | |
CF3CF2CF2CO2(CH2)3CH3 | |
MFCD00039240 | |
n-butyl heptafluorobutyrate, butyric acid, heptafluoro-, butyl ester, butyl heptafluorobutanoate, heptafluorobutyric acid, n-butyl ester, n-butylheptafluorobutyrate, butyl perfluorobutanoat, butyl perfluorobutyrate, butylheptafluorobutyrate, butyl heptafluorobutyrate, n-butyl perfluoro-n-butyrate | |
CCCCOC(=O)C(C(C(F)(F)F)(F)F)(F)F | |
270.147 | |
270.15 | |
n-Butyl heptafluorobutyrate |
1.296 | |
C8H9F7O2 | |
1.327 | |
1798327 | |
YDXXJZKOOOJVEE-UHFFFAOYSA-N | |
butyl 2,2,3,3,4,4,4-heptafluorobutanoate | |
137079 | |
97% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures