missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyloxycarbonyl-D-threonine, 98+%, Thermo Scientific™

Product Code. 11312586
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80384-27-6
Molecular Formula C12H15NO5
Molecular Weight (g/mol) 253.25
MDL Number MFCD00037808
InChI Key IPJUIRDNBFZGQN-WCBMZHEXSA-N
Synonym z-d-thr-oh, z-d-threonine, n-carbobenzoxy-d-threonine, 2r,3s-2-benzyloxy carbonyl amino-3-hydroxybutanoic acid, n-benzyloxycarbonyl-d-threonine, n-cbz-d-threonine, d-threonine, n-phenylmethoxy carbonyl, 2r,3s-3-hydroxy-2-phenylmethoxycarbonylamino butanoic acid, cbz-d-thr-oh
PubChem CID 853484
IUPAC Name (2R,3S)-3-hydroxy-2-(phenylmethoxycarbonylamino)butanoic acid
SMILES C[C@H](O)[C@@H](NC(=O)OCC1=CC=CC=C1)C(O)=O

Specifications

Melting Point 100°C to 105°C
Quantity 1 g
Beilstein 4695777
Optical Rotation +5.5° (c=2 in Acetic Acid)
Formula Weight 253.26
Percent Purity ≥98%
Chemical Name or Material N-Benzyloxycarbonyl-D-threonine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'