missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N-Acetyl-L-phenylalanine, 99%

20.70 € - 188.00 €

Chemical Identifiers

CAS 2018-61-3
Molecular Formula C11H13NO3
Molecular Weight (g/mol) 207.23
MDL Number MFCD00063158
InChI Key CBQJSKKFNMDLON-JTQLQIEISA-N
Synonym n-acetyl-l-phenylalanine, ac-phe-oh, acetyl-l-phenylalanine, acetylphenylalanine, l-n-acetylphenylalanine, s-2-acetamido-3-phenylpropanoic acid, phenylalanine, n-acetyl, n-acetyl-3-phenyl-l-alanine, unii-np5bt39467, 2s-2-acetamido-3-phenylpropanoic acid
PubChem CID 74839
ChEBI CHEBI:16259
IUPAC Name (2S)-2-acetamido-3-phenylpropanoic acid
SMILES CC(=O)N[C@@H](CC1=CC=CC=C1)C(O)=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11420278
View Documents Promotion Details
Thermo Scientific Chemicals
B23812.03
1 g
20.70 €
1g
Please to purchase this item. Need a web account? Register with us today!
11430278
View Documents Promotion Details
Thermo Scientific Chemicals
B23812.06
5 g
49.70 €
5g
Please to purchase this item. Need a web account? Register with us today!
11440278
View Documents Promotion Details
Thermo Scientific Chemicals
B23812.14
25 g
188.00 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

It is an essential amino acid produced for medical, feed, and nutritional applications such as in the preparation of aspartame. It is applied as an antidepressant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an essential amino acid produced for medical, feed, and nutritional applications such as in the preparation of aspartame. It is applied as an antidepressant.

Solubility
Solubility in methanol (almost transparency). Soluble in acetone and ethanol (20 mg/ ml).

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Chemical Identifiers

2018-61-3
207.23
CBQJSKKFNMDLON-JTQLQIEISA-N
74839
(2S)-2-acetamido-3-phenylpropanoic acid
C11H13NO3
MFCD00063158
n-acetyl-l-phenylalanine, ac-phe-oh, acetyl-l-phenylalanine, acetylphenylalanine, l-n-acetylphenylalanine, s-2-acetamido-3-phenylpropanoic acid, phenylalanine, n-acetyl, n-acetyl-3-phenyl-l-alanine, unii-np5bt39467, 2s-2-acetamido-3-phenylpropanoic acid
CHEBI:16259
CC(=O)N[C@@H](CC1=CC=CC=C1)C(O)=O

Specifications

166°C to 170°C
C11H13NO3
n-acetyl-l-phenylalanine, ac-phe-oh, acetyl-l-phenylalanine, acetylphenylalanine, l-n-acetylphenylalanine, s-2-acetamido-3-phenylpropanoic acid, phenylalanine, n-acetyl, n-acetyl-3-phenyl-l-alanine, unii-np5bt39467, 2s-2-acetamido-3-phenylpropanoic acid
CC(=O)N[C@@H](CC1=CC=CC=C1)C(O)=O
207.23
CHEBI:16259
99%
N-Acetyl-L-phenylalanine
2018-61-3
MFCD00063158
CBQJSKKFNMDLON-JTQLQIEISA-N
(2S)-2-acetamido-3-phenylpropanoic acid
74839
207.23
+40° (c=1 in Methanol)
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 217-959-8

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only