Learn More
N-(3-Aminopropyl)imidazole, 98%, Thermo Scientific Chemicals
44.42 € - 145.73 €
Chemical Identifiers
CAS | 5036-48-6 |
---|---|
Molecular Formula | C6H11N3 |
Molecular Weight (g/mol) | 125.18 |
MDL Number | MFCD00009819 |
InChI Key | KDHWOCLBMVSZPG-UHFFFAOYSA-N |
Synonym | 1-3-aminopropyl imidazole, n-3-aminopropyl imidazole, 1h-imidazole-1-propanamine, n-3-aminopropyl-imidazole, 3-1h-imidazol-1-yl propan-1-amine, 3-imidazol-1-ylpropylamine, 3-1h-imidazol-1-yl-1-propanamine, 1-3-aminopropyl-imidazole, 3-imidazol-1-yl propylamine, 3-imidazolylpropylamine |
PubChem CID | 78736 |
IUPAC Name | 3-imidazol-1-ylpropan-1-amine |
SMILES | NCCCN1C=CN=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10584721
|
Thermo Scientific Acros
351940500 |
50 g | Glass bottle |
44.42 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10439671
|
Thermo Scientific Acros
351942500 |
250 g | Glass bottle |
145.73 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
5036-48-6 | |
125.18 | |
KDHWOCLBMVSZPG-UHFFFAOYSA-N | |
78736 | |
NCCCN1C=CN=C1 |
C6H11N3 | |
MFCD00009819 | |
1-3-aminopropyl imidazole, n-3-aminopropyl imidazole, 1h-imidazole-1-propanamine, n-3-aminopropyl-imidazole, 3-1h-imidazol-1-yl propan-1-amine, 3-imidazol-1-ylpropylamine, 3-1h-imidazol-1-yl-1-propanamine, 1-3-aminopropyl-imidazole, 3-imidazol-1-yl propylamine, 3-imidazolylpropylamine | |
3-imidazol-1-ylpropan-1-amine |
Specifications
5036-48-6 | |
100.0 | |
Colorless | |
296.0°C | |
Authentic | |
C6H11N3 | |
MFCD00009819 | |
1-3-aminopropyl imidazole, n-3-aminopropyl imidazole, 1h-imidazole-1-propanamine, n-3-aminopropyl-imidazole, 3-1h-imidazol-1-yl propan-1-amine, 3-imidazol-1-ylpropylamine, 3-1h-imidazol-1-yl-1-propanamine, 1-3-aminopropyl-imidazole, 3-imidazol-1-yl propylamine, 3-imidazolylpropylamine | |
KDHWOCLBMVSZPG-UHFFFAOYSA-N | |
3-imidazol-1-ylpropan-1-amine | |
78736 | |
98% | |
N-(3-Aminopropyl)imidazole, 98% |
97.5 | |
-68.0°C | |
1.0620g/mL | |
154°C | |
97.5% min. (GC) | |
1.5175 to 1.5195 | |
1.062 | |
Solubility in water: miscible. | |
NCCCN1C=CN=C1 | |
125.18 | |
125.17 | |
Liquid |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful to aquatic life.
Harmful to aquatic life with long lasting effects.
GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 225-730-9
missing translation for 'tsca' : TSCA
- N-(3-Aminopropyl)imidazole
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H400-Very toxic to aquatic life.
- H314-Causes severe skin burns and eye damage.
- H412-Harmful to aquatic life with long lasting effects.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only