missing translation for 'onlineSavingsMsg'
Learn More

N-(3-Acetyl-4-hydroxyphenyl)butyramide, 97%, Thermo Scientific Chemicals

Product Code. 11472958
Click to view available options
Quantity:
50 g
250 g
1000 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40188-45-2
Molecular Formula C12H15NO3
Molecular Weight (g/mol) 221.26
MDL Number MFCD00798556
InChI Key FGWZEOPEZISTTR-UHFFFAOYSA-N
Synonym 2-acetyl-4-butyramidophenol, n-3-acetyl-4-hydroxyphenyl butanamide, n-3-acetyl-4-hydroxyphenyl butyramide, ac-hophe-oh, unii-14qjb65r4v, 3'-acetyl-4'-hydroxybutyranilide, butanamide, n-3-acetyl-4-hydroxyphenyl, 5'-butyramido-2'-hydroxyacetophenone, acebutolol impurity c, n-3-acetyl-4-hydroxyphenyl butanamid
PubChem CID 736331
IUPAC Name N-(3-acetyl-4-hydroxyphenyl)butanamide
SMILES CCCC(=O)NC1=CC=C(O)C(=C1)C(C)=O

Specifications

Melting Point 124°C to 126°C
Quantity 1000 g
Formula Weight 221.26
Percent Purity 97%
Chemical Name or Material N-(3-Acetyl-4-hydroxyphenyl)butyramide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'