missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYH9 Mouse anti-Human, Mouse, Porcine, Clone: 5D9D2, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 60233-1-IG-150UL
This item is not returnable.
View return policy
Description
TMP21 is a ubiquitously expressed protein that is involved in vesicular targeting and protein transport. More recent experiments have shown that it is also a component in the presenilin complex and modulates the -gamma-secretase but not the epsilon-secretase cleavage activity of the amyloid precursor protein. The presenilin complex is composed of the proteins APH1, nicastrin, and PEN2 in addition to presenilin-1. Together, these proteins cleave the amyloid precursor protein at what is known as the -gamma- and epsilon-sites and can lead to the accumulation of the Abeta cleavage product that is associated with Alzheimer′s disease. Co-immunoprecipitation experiments using antibodies against these proteins also yielded TMP21 indicating that TMP21 may play a role in the regulation of this complex. Suppression of TMP21 expression by siRNA in transfected cells caused increased -gamma-secretase activity but not epsilon-secretase activity, and increased Abeta production, demonstrating that TMP21 can modulate -gamma-secretase activity.Specifications
MYH9 | |
Monoclonal | |
0.61 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P35579, Q8VDD5 | |
Myh9 | |
MYH9 Fusion Protein Ag1612 | |
150 μL | |
Primary | |
Human, Mouse, Porcine | |
Antibody | |
IgG1 |
Immunocytochemistry, Immunofluorescence, Western Blot | |
5D9D2 | |
Unconjugated | |
MYH9 | |
DFNA17, EPSTS, FTNS, MHA, MYH9, Myosin 9, Myosin heavy chain 9 | |
Mouse | |
Protein G | |
RUO | |
17886, 396904, 4627 | |
-20°C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction