missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYH6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 22281-1-AP-150UL
This item is not returnable.
View return policy
Description
Myosin binding protein C (MYBPC) is a component of the thick filament of striated muscle, with the fast-type isoform designated MYBPC2. Both the fast-type and slow-type MYBPC protein contains seven immunoglobulin C2 motifs and three fibronectin type-III repeats. MYBPC2 is typically required for strong contractions and functions under anaerobic conditions. It is more similar to the cardiac isoform (MYBPC3) than to the slow-type isoform (MYBPC1) in terms of pCa50-indexed force development, length-independent cooperativity and length dependent activation. It has been suggested that in cardiac and fast muscle MYBPC2 contributes to an internal load, possibly by binding to actin via its N-terminal region.Specifications
MYH6 | |
Polyclonal | |
Unconjugated | |
MYH6 | |
alpha MHC, ASD3, MYH6, MYHC, MyHC alpha, MYHCA, Myosin 6, Myosin heavy chain 6 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
17888, 29556, 4624 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.43 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P02563, P13533, Q02566 | |
Myh6 | |
Peptide | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |