missing translation for 'onlineSavingsMsg'
Learn More

ZVG Zellstoff Vertrieb™ Multiclean Plus Cloth Rolls

Clean using ZVG Zellstoff Vertrieb™ Multiclean Plus Cloth Rolls for a high degree of efficiency and safety A new manufacturing process makes these cloths even stronger and more absorbent.

38.05 €

Specifications

Product Type Multiclean Plus Cloth Roll
Length (Metric) 380 mm
Color Blue
View More Specs

Products 1
Product Code Brand Ply Width (Metric) Price Quantity & Availability  
Product Code Brand Ply Width (Metric) Price Quantity & Availability  
10116881
missing translation for 'viewDocuments'
ZVG Zellstoff Vertrieb™
11224-00
2-ply 240 mm
38.05 €
2 rolls
Estimated Shipment: 16-01-2025
to see stock.
Add to basket
 
Description

Description

  • Blue
  • 2-ply and 3-ply
  • High volume
Specifications

Specifications

Multiclean Plus Cloth Roll
380 mm
Blue
Videos
SDS
Documents

Documents

Certificates
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.