missing translation for 'onlineSavingsMsg'
Learn More

Sampling Systems™ Multi Purpose V Spatula

Sampling Systems™ Multi Purpose V Spatula is ideal for surface sampling of powders and core sampling of viscous material like cheese, pastes and gels.

Brand:  Sampling Systems™ 1095S-150

Product Code. 15869211

  • 85.00 € / Each
Estimated Shipment: 20-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Made from high quality 316L stainless steel
  • Polished finish
  • Fully welded, crevice free construction
  • V Shape allows for easy discharge of powders into containers or bags
Specifications

Specifications

Stainless Steel (316L)
230 mm
85 mm
35 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.