Learn More
Sampling Systems™ Multi Dipper 2
Maintain a closed sample collection bottle until the operator uses the control chain to remove the stoppers. Sampling Systems™ Multi Dipper 2 is the ideal solution for taking liquid samples from different levels.
Brand:  Sampling Systems™ 5045A-1500
 View more versions of this product
Product Code. 15839341
  • 710.00 € / Each
Estimated Shipment: 17-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Sampler frame, control chains, plugs and plug shackles are constructed of Stainless Steel
  • Sampler bottle are constructed of High Density Polyethylene (HDPE)
  • Ideal solution for taking liquid samples from different levels
  • The sample collection bottles remain closed until the operator uses the control chain to remove the stoppers
Specifications
Specifications
Dipper
3 HDPE Sampling Bottles
Liquid Sampling
Stainless Steel (316L)
3 x 250 mL
1500 mm