missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MUC20 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13380-1-AP-20UL
This item is not returnable.
View return policy
Description
Mucins are a family of high molecular weight, heavily glycosylated proteins (glycoconjugates) produced by epithelial tissues in most organisms of Kingdom Animalia. Mucins′ key characteristic is their ability to form gels; therefore they are a key component in most gel-like secretions, serving functions from lubrication to cell signalling to forming chemical barriers. They often take an inhibitory role. Some mucins are associated with controlling mineralization, including nacre formation in mollusks, calcification in echinoderms and bone formation in vertebrates. They bind to pathogens as part of the immune system. Overexpression of the mucin proteins, especially MUC1, is associated with many types of cancer.Specifications
MUC20 | |
Polyclonal | |
Unconjugated | |
MUC20 | |
KIAA1359, MUC 20, MUC20, Mucin 20 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
200958 | |
-20°C | |
Liquid |
Western Blot, Immunoprecipitation | |
0.31 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8N307 | |
MUC20 | |
MUC20 Fusion Protein Ag4205 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |