Learn More
MTCH2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 16888-1-AP-150UL
 View more versions of this product
Product Code. 16806654
  • 410.00 € / 150µL
Explore more special offers
Description
Description
MTCH2 is a member of the mitochondrial carrier protein family that catalyze the exchange of substrates across the inner mitochondrial membrane and is targeted by Bid, a pro-apoptotic Bcl-2 family member, in response to apoptotic signals, suggesting that MTCH2 may play a key role in the mitochondrial apoptosis pathway. Apoptosis plays a major role in normal organism development, tissue homeostasis, and removal of damaged cells. Disruption of this process has been implicated in a variety of diseases such as cancer. The Bcl-2 family of proteins is comprised of critical regulators of apoptosis that can be divided into two classes: those that inhibit apoptosis and those that promote cell death.
Specifications
Specifications
MTCH2
Polyclonal
Unconjugated
MTCH2
HSPC032, MIMP, MTCH2
Rabbit
Antigen Affinity Chromatography
RUO
23788, 56428
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.24 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q791V5, Q9Y6C9
Mtch2
MTCH2 Fusion Protein Ag10399
150 μL
Primary
Human, Mouse