missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTAP Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00 € - 410.00 €
Specifications
Antigen | MTAP |
---|---|
Concentration | 367 μg/mL |
Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16818583
|
Proteintech
11475-1-AP-20UL |
20 μL |
145.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16808583
|
Proteintech
11475-1-AP-150UL |
150 μL |
410.00 €
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The MTAP antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human MTAP. This antibody recognizes human, rat, mouse antigen. The MTAP antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
MTAP | |
Western Blot, Immunohistochemistry, Immunofluorescence | |
Unconjugated | |
Human, Rat, Mouse | |
IgG |
367 μg/mL | |
Polyclonal | |
Rabbit | |
4507 | |
Primary |