missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MSH3 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
140.00 € - 399.00 €
Specifications
Antigen | MSH3 |
---|---|
Concentration | 0.27 mg/mL |
Applications | Western Blot, Immunoprecipitation |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16851915
|
Proteintech
22393-1-AP-20UL |
20 μL |
140.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16841915
|
Proteintech
22393-1-AP-150UL |
150 μL |
399.00 €
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PUS1 converts uridine into pseudouridine after the nucleotide has been incorporated into RNA. Pseudouridine may have a functional role in tRNAs and may assist in the peptidyl transfer reaction of rRNAs.Specifications
MSH3 | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Human | |
P20585 | |
4437 | |
MSH3 Fusion Protein Ag18060 | |
Primary | |
-20°C | |
MSH3 |
0.27 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Divergent upstream protein, DUC1, DUG, DUP, hMSH3, Mismatch repair protein 1, MRP1, MSH3, mutS homolog 3 (E. coli) | |
MSH3 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |