missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MPP3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14650-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene product is a member of a family of membrane-associated proteins termed MAGUKs (membrane-associated guanylate kinase homologs). MAGUKs interact with the cytoskeleton and regulate cell proliferation, signaling pathways, and intracellular junctions. This protein contains a conserved sequence, called the SH3 (src homology 3) motif, found in several other proteins that associate with the cytoskeleton and are suspected to play important roles in signal transduction. Alternatively spliced transcript variants have been identified. One transcript variant is experimentally supported, but it doesn′t encode a protein.Specifications
MPP3 | |
Polyclonal | |
Unconjugated | |
MPP3 | |
Discs large homolog 3, DLG3, MAGUK p55 subfamily member 3, MPP3, Protein MPP3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
114202, 13384, 4356 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O88910, O88954, Q13368 | |
MPP3 | |
MPP3 Fusion Protein Ag6246 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction