Learn More
Welch Ilmvac™ MPC 301 Z Vacuum Pump
Use these pumps for chemical duty applications. Welch Ilmvac™ MPC 301 Z Vacuum Pumps use PTFE and other fluorinated plastics for the wetted parts to allow aggressive solvent and acid vapors to be pumped.
Brand: Welch Ilmvac™ 412722
This item is not returnable.
View return policy
Description
Use these pumps for chemical duty applications. Welch Ilmvac™ MPC 301 Z Vacuum Pumps use PTFE and other fluorinated plastics for the wetted parts to allow aggressive solvent and acid vapors to be pumped.
- Low weight and excellent ergonomics
- Proven diaphragm technology offers a double benefit to the user
- ON/OFF switch and internal protective thermal switch for the motor, mains cable and plug
- Carry handle
- Vibration isolating feet
- MP models come with and exhaust silencer
- MPC models come with a gas ballast valve
- analytically pure, oil free vacuum
- User friendly
- Light weight, rugged design
- Designed for permanent operation
- Maintenance-free drive system and proven long diaphragm life
- Minimal operation vibration
- Wide vacuum and flow range to match application
- Superior chemical resistance (MPC models)
Specifications
29.7 in.Hg | |
180 w | |
41 L/min. | |
IP 54 | |
8 mbar |
23 x 26.5 x 16.9 cm | |
11.2 kg | |
Vacuum Pump | |
Chemical Duty |