missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Transport Factors
752.00 € - 776.98 €
Specifications
Content And Storage | -20°C |
---|---|
Product Type | Transferrin |
Quantity | 1 g |
Source | Pooled Human Plasma |
Description
Transport factors are proteins which enable small metabolites to be carried in usable form to cells. Transferrin binds iron and makes it available to cells in a recognizable form.
†Contains <1.0% iron.
Specifications
-20°C | |
Transferrin | |
1 g | |
Pooled Human Plasma |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
MP Biomedicals™ Transport Factors