missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Lysozyme from Chicken Egg White
2X crystallized, salt-free and soluble
Brand: MP Biomedicals™ 0210083410
This item is not returnable.
View return policy
Description
Activity: >9000 units/mg protein. Unit Definition: One unit will cause a decrease in A450 of 0.001 per minute at pH 6.24 and 25°C using Micrococcus lysodeikticus as substrate.
- Lyophilized
- Synonyms: Muramidase, Mucopeptide N-acetylmuramoylhydrolase
- CAS: 9001-63-2
Specifications
2X | |
11 | |
0°C | |
10 g | |
14388 | |
Crystallized, Lyophilized |
White | |
Lysing E. coli and Streptomycetes for extraction purposes such as extracting group specific Antigen | |
Chicken egg white | |
Lysozyme | |
Surface-active reagents such as Dodecyl Sulfate, Alcohols and Fatty Acids, Imidazole and Indole derivatives are inhibitors via formation of change-transfer complexes |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Lysozyme
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Skin sensitiser Category 1
Hazard Statement
- H302-Harmful if swallowed.
- H317-May cause an allergic skin reaction.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P321-Specific treatment (see on this label).
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Lysozyme
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Research Use Only
Spot an opportunity for improvement?Share a Content Correction