missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Guinea Pig Complement, 15mL
Brand: MP Biomedicals™ 08642836
This item is not returnable.
View return policy
Description
Specifications
Immunoassay | |
15 mL | |
2° to 8°C | |
Liquid |
Guinea Pig Complement | |
Guinea Pig | |
Guinea Pig |