missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Glucose Oxidase

From Aspergillus niger

Brand:  MP Biomedicals™ 0219519690

519.19 EUR valid until 2025-12-31
Use promo code "24274" to get your promotional price.



 View more versions of this product

Product Code. 11411102

  • 947.00 € / 500ku

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Type X. Activity: >100U/mg. Unit Definition: one unit oxidizes 1.0μmol of β-D-glucose to D-gluconic acid and H2O2 per minute at 35°C, pH 5.1.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

>100 U/mg
4 to 7
-20°C
500 kU
≈160000
Lyophilized Powder
Yellow
Glucose Oxidase can be utilized in the enzymatic determination of D-glucose in solution
Aspergillus niger
Glucose Oxidase
Ag+, Hg2+, Cu2+ , 4-chloromercuribenzoate, 50% D-arabinose
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Glucose Oxidase

Signal Word
  • Danger

Hazard Category
  • Respiratory sensitiser Category 1

Hazard Statement
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : Glucose Oxidase

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.