missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Geneclean™ Turbo Kit

Used for purification of DNA fragments of sizes 0.1 kb to 300 kb from agarose gels, PCR reactions and other enzymatic solutions. MP Biomedicals™ Geneclean™ Turbo Kit can be used in enzymatic reactions, conventional or automated sequencing, cloning and microarray analysis.

171.00 € - 605.00 €

Specifications

Content And Storage Room Temperature
View More Specs

Products 3
Product Code Brand No. of Reactions Price Quantity & Availability  
Product Code Brand No. of Reactions Price Quantity & Availability  
11393119
View Documents Promotion Details
MP Biomedicals™
111102200
50 Preps
171.00 €
50 preps
Please to purchase this item. Need a web account? Register with us today!
15122185
View Documents Promotion Details
MP Biomedicals™
111102400
100 Preps
259.00 €
1 set
Please to purchase this item. Need a web account? Register with us today!
11313129
View Documents Promotion Details
MP Biomedicals™
111102600
300 Preps
605.00 €
300 preps
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Process with either vacuum or centrifugation and save time with patented Turbo filter design
  • Patented GeneClean™ technology simplifies the process of purifying DNA into three easy steps: Bind, Wash, and Elute
  • Ethanol precipitation is never required, and purified DNA is immediately ready for a wide variety of downstream applications
Specifications

Specifications

Room Temperature
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • geneclean turbo kit

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H412-Harmful to aquatic life with long lasting effects.
  • EUH032-Contact with acids liberates very toxic gas.

Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : guanidine thiocyanate

SDS
Documents

Documents

Product Certifications

Research Use Only (RUO)