Learn More
MP Biomedicals™ Dispase
From Bacillus polymyxa
Brand: MP Biomedicals™ 0219502225
Description
Microbial metalloenzyme used in tissue dissociation (primary cell cultures) and subcultivation (secondary cell cultures). Extremely stable. Activity: 6U/mg. Unit Definition: One unit is the amount of enzyme which liberates 1.0μmole of Tyr per minute at 37°C, pH 7.5. MW: 35.9kDa. Lyophilized.
Ref.: Akimoto, Y., et al., J. Histochem. Cytochem., 40: 1129 (1992).
Specifications
Tan | |
Protein sequencing | |
Bacillus polymyxa | |
Dispase | |
EDTA, EGTA, Hg2+, other heavy Metals |
6 to 8.5 | |
4°C | |
25 U | |
35.9 | |
Lyophilized Powder |
Safety and Handling
- Danger
Hazard Category
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H335-May cause respiratory irritation.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H319-Causes serious eye irritation.
Precautionary Statement
- P501-Dispose of contents/container to in accordance with local/regional/national/international regulations.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P405-Store locked up.
- P103-Read label before use.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.