missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Bovine Transferrin, (HOLO)

Suitable for regulating iron levels in cell culture

Brand:  MP Biomedicals™ 0215233580

 View more versions of this product

Product Code. 17512726

  • 126.00 € / 100mg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Transferrin is a 77kDa plasma glycoprotein with the capacity to bind ferric ions and as such is a source of bio-available iron. The product is frequently used as a nutrient in cell and microbial culture and as a media supplement in the production of bio-pharmaceutical products. Transferrin has also been shown to function as a factor in the growth stimulation of many cell types, such as granulocytes, macrophages and endothelial cells. Chromatographically purified.

Physical characteristics:

  • Appearance: Reddish-brown powder
  • Purity: ≥95% w/w
  • Iron: ≥1200 mg/g protein
  • Moisture: ≤5%
  • pH (3%): 6.0 to 8.0
  • Solubility: Dissolves in <20 min.
Microbial evlauation and control:
  • Bioburden: ≤100cfu/g
  • Endotoxin: ≤1 EU/mg
  • Mycoplasma: Not detected
  • Virus: Not detected
Certifications: ISO9000, cGMP, EDQM

TRUSTED_SUSTAINABILITY
Specifications

Specifications

6 to 8
Bovine Serum
Bovine Transferrin (HOLO)
100 mg
4°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers