missing translation for 'onlineSavingsMsg'
Learn More

Proteintech™ Mouse CXCL12/SDF-1 ELISA Kit

Mouse CXCL12/SDF-1 ELISA Kit

Brand:  Proteintech™ KE10049-96T


Product Code. 16801646

  • 269.00 € / 96 tests
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • The chemokine stroma-derived factor (SDF-1/CXCL12) plays multiple roles in tumor
Specifications

Specifications

BC006640
25.8 pg/mL
Sandwich ELISA, Double Antibody
Plasma, Serum
Mouse
Mouse
<10%
Blue ice
4.5 hr.
62.5 to 4000 pg/mL
ISO 13485
Coated ELISA Kit
DAB
20315
Solid
<10%
-20°C to +4°C
P40224
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 96TEST Human CRP ELISA Kit

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P234-Keep only in original container.
  • P264-Wash skin thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P330-Rinse mouth.
  • P362-Take off contaminated clothing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : Sulphuric Acid, Tetramethylbenzidine

SDS
Documents

Documents

Certificates
Special Offers

Special Offers