Learn More
MOPSO, 98%, Thermo Scientific Chemicals
Biological buffers, pH range 6.2 to 7.6
23.50 € - 843.00 €
Chemical Identifiers
CAS | 68399-77-9 |
---|---|
Molecular Formula | C7H15NO5S |
Molecular Weight (g/mol) | 225.259 |
MDL Number | MFCD00009629 |
InChI Key | NUFBIAUZAMHTSP-UHFFFAOYSA-N |
Synonym | mopso, 2-hydroxy-3-morpholinopropanesulfonic acid, 2-hydroxy-3-morpholinopropane-1-sulfonic acid, 2-hydroxy-4-morpholinepropanesulphonic acid, 3-morpholino-2-hydroxypropanesulfonic acid, 3-n-morpholino-2-hydroxypropanesulfonic acid, 2-hydroxy-3-morpholin-4-yl propane-1-sulfonic acid, beta-hydroxy-4-morpholinepropanesulfonic acid, 3-4-morpholino-2-hydroxypropanesulfonic acid, 4-morpholinepropanesulfonic acid, .beta.-hydroxy |
PubChem CID | 109333 |
IUPAC Name | 2-hydroxy-3-morpholin-4-ylpropane-1-sulfonic acid |
SMILES | C1COCCN1CC(CS(=O)(=O)O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11329628
|
Thermo Scientific Alfa Aesar
044135.09 |
10 g | |||||||
11319628
|
Thermo Scientific Alfa Aesar
044135.18 |
50 g |
74.90 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11309628
|
Thermo Scientific Alfa Aesar
044135.30 |
250 g |
297.00 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11339628
|
Thermo Scientific Alfa Aesar
044135.A1 |
1 kg |
843.00 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is an important raw material used intermediate used in pharmaceuticals, organic synthesis, dye stuffs and agrochemicals. Also used as biological buffers, pH range 6.2-7.6.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
68399-77-9 | |
225.259 | |
NUFBIAUZAMHTSP-UHFFFAOYSA-N | |
109333 | |
C1COCCN1CC(CS(=O)(=O)O)O |
C7H15NO5S | |
MFCD00009629 | |
mopso, 2-hydroxy-3-morpholinopropanesulfonic acid, 2-hydroxy-3-morpholinopropane-1-sulfonic acid, 2-hydroxy-4-morpholinepropanesulphonic acid, 3-morpholino-2-hydroxypropanesulfonic acid, 3-n-morpholino-2-hydroxypropanesulfonic acid, 2-hydroxy-3-morpholin-4-yl propane-1-sulfonic acid, beta-hydroxy-4-morpholinepropanesulfonic acid, 3-4-morpholino-2-hydroxypropanesulfonic acid, 4-morpholinepropanesulfonic acid, .beta.-hydroxy | |
2-hydroxy-3-morpholin-4-ylpropane-1-sulfonic acid |
Specifications
68399-77-9 | |
Powder | |
MFCD00009629 | |
Water solubility under desired conditions ca.112,6g/L at 20°C. | |
C1COCCN1CC(CS(=O)(=O)O)O | |
225.259 | |
225.27 | |
MOPSO |
276°C to 279°C | |
C7H15NO5S | |
mopso, 2-hydroxy-3-morpholinopropanesulfonic acid, 2-hydroxy-3-morpholinopropane-1-sulfonic acid, 2-hydroxy-4-morpholinepropanesulphonic acid, 3-morpholino-2-hydroxypropanesulfonic acid, 3-n-morpholino-2-hydroxypropanesulfonic acid, 2-hydroxy-3-morpholin-4-yl propane-1-sulfonic acid, beta-hydroxy-4-morpholinepropanesulfonic acid, 3-4-morpholino-2-hydroxypropanesulfonic acid, 4-morpholinepropanesulfonic acid, .beta.-hydroxy | |
NUFBIAUZAMHTSP-UHFFFAOYSA-N | |
2-hydroxy-3-morpholin-4-ylpropane-1-sulfonic acid | |
109333 | |
98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 269-989-6
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only