missing translation for 'onlineSavingsMsg'
Learn More

MOPS, MP Biomedicals™

Product Code. 11483610
Click to view available options
Quantity:
10 g
100 g
250 g
1 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 1132-61-2
Molecular Formula C7H15NO4S
Molecular Weight (g/mol) 209.26
MDL Number MFCD00006183
InChI Key DVLFYONBTKHTER-UHFFFAOYSA-N
Synonym 3-(N-morpholino)propanesulfonic acid, 4-morpholinepropanesulfonic acid
PubChem CID 70807
ChEBI CHEBI:44115
SMILES [O-]S(=O)(=O)CCC[NH+]1CCOCC1

Specifications

Color White
Physical Form Powder
Quantity 100 g
Assay Percent Range 100.4%
IUPAC Name 4-(3-sulfonatopropyl)morpholin-4-ium
Formula Weight 209.3
Chemical Name or Material MOPS
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.