missing translation for 'onlineSavingsMsg'
Learn More

MOPS-buffered saline (5X), pH 7.0, Thermo Scientific™

Product Code. 15456129
Click to view available options
Quantity:
250 mL
500 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1132-61-2
Molecular Formula C7H15NO4S
Molecular Weight (g/mol) 209.26
MDL Number MFCD00006183
InChI Key DVLFYONBTKHTER-UHFFFAOYSA-N
PubChem CID 70807
ChEBI CHEBI:44115
IUPAC Name 3-morpholin-4-ylpropane-1-sulfonic acid
SMILES [O-]S(=O)(=O)CCC[NH+]1CCOCC1

Specifications

Concentration 5X
pH 7.0
Physical Form Liquid
Quantity 500 mL
Chemical Name or Material MOPS-buffered saline
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'