Learn More
Thermo Scientific Chemicals MOPS, 1.0M buffer soln., pH 6.5
94.17 € - 142.71 €
Chemical Identifiers
CAS | 1132-61-2 |
---|---|
Molecular Formula | C7H15NO4S |
Molecular Weight (g/mol) | 209.26 |
MDL Number | MFCD00006183 |
InChI Key | DVLFYONBTKHTER-UHFFFAOYSA-N |
PubChem CID | 70807 |
ChEBI | CHEBI:44115 |
IUPAC Name | 3-morpholin-4-ylpropane-1-sulfonic acid |
SMILES | [O-]S(=O)(=O)CCC[NH+]1CCOCC1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15484309
|
Thermo Scientific Chemicals
J61797.AK |
250 mL |
94.17 €
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15494309
|
Thermo Scientific Chemicals
J61797.AP |
500 mL |
142.71 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MOPS is frequently used as a buffering agent in biology and biochemistry. It used widely for polyacrylamide gel electrophoresis. It is used as a morpholino propanesulfonic acid buffer.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsMOPS is frequently used as a buffering agent in biology and biochemistry. It used widely for polyacrylamide gel electrophoresis. It is used as a morpholino propanesulfonic acid buffer.
Solubility
Soluble in water.
Notes
Usage above 20 mM in mammalian cell culture work is not recommended. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Strong oxidizing agents, Strong bases.
Chemical Identifiers
1132-61-2 | |
209.26 | |
DVLFYONBTKHTER-UHFFFAOYSA-N | |
CHEBI:44115 | |
[O-]S(=O)(=O)CCC[NH+]1CCOCC1 |
C7H15NO4S | |
MFCD00006183 | |
70807 | |
3-morpholin-4-ylpropane-1-sulfonic acid |
Specifications
1132-61-2 | |
1.0 M | |
Liquid | |
Odorless | |
Soluble in water. | |
[O-]S(=O)(=O)CCC[NH+]1CCOCC1 | |
209.26 | |
CHEBI:44115 |
Colorless | |
6.5 | |
C7H15NO4S | |
MFCD00006183 | |
DVLFYONBTKHTER-UHFFFAOYSA-N | |
3-morpholin-4-ylpropane-1-sulfonic acid | |
70807 | |
MOPS buffer soln. |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.