missing translation for 'onlineSavingsMsg'
Learn More

MOPS, 0.5M buffer soln., pH 8.5, Thermo Scientific™

42.70 € - 64.00 €

Chemical Identifiers

CAS 1132-61-2
Molecular Formula C7H15NO4S
Molecular Weight (g/mol) 209.26
MDL Number MFCD00006183
InChI Key DVLFYONBTKHTER-UHFFFAOYSA-N
PubChem CID 70807
ChEBI CHEBI:44115
IUPAC Name 3-morpholin-4-ylpropane-1-sulfonic acid
SMILES [O-]S(=O)(=O)CCC[NH+]1CCOCC1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15404769
SDS View Documents
Thermo Scientific Alfa Aesar
J62368.AE
100 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15414769
SDS View Documents
Thermo Scientific Alfa Aesar
J62368.AK
250 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
MOPS is frequently used as a buffering agent in biology and biochemistry. It used widely for polyacrylamide gel electrophoresis. It is used as a A morpholino propanesulfonic acid buffer.

Solubility
Soluble in water.

Notes
Usage above 20 mM in mammalian cell culture work is not recommended. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Strong oxidizing agents, Strong bases.
Specifications
Show More
Videos
Safety and Handling

Safety and Handling

P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362

H315-H319-H335

missing translation for 'tsca' : Yes

Recommended Storage : Keep cold

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
MOPS, 0.5M buffer soln., pH 8.5, Thermo Scientific™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.