Learn More
Micronova™ Mop Cover Controlled Environment
The MegaTex™ T-Mop cover is ideal for abrasion resistant cleanroom and production area walls and floors. For use with the QDT-18 and SSU series handles
Brand:  Micronova™ T20-18
Product Code. 13177686
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
MegaTex cover, with thin yellow sponge, 457mm TMop